missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aroclor 1260 in Methanol 1000μg/mL, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Packaging | Ampoule |
| CAS | 11096-82-5 |
| Molecular Formula | C12H4Cl6 |
| Concentration | 1000 μg/mL |
| InChI Key | BTAGRXWGMYTPBY-UHFFFAOYSA-N |
| Type | Organic Single-component Standard Solution |
| SMILES | C1=CC(=C(C(=C1C2=C(C(=C(C=C2)Cl)Cl)Cl)Cl)Cl)Cl |
| IUPAC Name | 1,2,3-trichloro-4-(2,3,4-trichlorophenyl)benzene |
| Solvent or Matrix | Methanol |
| Molecular Weight (g/mol) | 360.864 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction