missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-3,5-dimethylphenyl-N-methylcarbamate in Methanol 100μg/mL, Fisher Chemical™
Click to view available options
Quantity:
10 mL
Packaging:
Ampoule
Specifications
Concentration | 100μg/mL |
CAS | 672-99-1 |
Packaging | Ampoule |
Quantity | 10 mL |
Type | Organic Single-component Standard Solution |
Molecular Formula | C10H12BrNO2 |
MDL Number | MFCD00027941 |
InChI Key | ZOZFMTULOYRWEL-UHFFFAOYSA-N |
SMILES | CNC(=O)OC1=CC(C)=C(Br)C(C)=C1 |
IUPAC Name | (4-bromo-3,5-dimethylphenyl) N-methylcarbamate |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'