Learn More
Metabolite 2,4'-DDE in Methanol 100μg/mL, Fisher Chemical™
Product Code. 12952634 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. 12952634
Brand: Fisher Chemical P815720
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Packaging Ampoule(s)
CAS 3424-82-6
Molecular Formula C14H8Cl4
Concentration 100 μg/mL
InChI Key ZDYJWDIWLRZXDB-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)C(=C(Cl)Cl)C2=CC=C(C=C2)Cl)Cl
IUPAC Name 1-chloro-2-[2,2-dichloro-1-(4-chlorophenyl)ethenyl]benzene
Solvent or Matrix Methanol
Molecular Weight (g/mol) 318.018
PubChem CID 246598
Quantity 1 mL
ChEBI CHEBI:34211
Chemical Name or Material 2,4'-DDE