missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress meta-iodoHoechst 33258
Specifications
Specifications
| Content And Storage | 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light) |
| Quantity | 100 mg |
| Shipping Condition | Room temperature |
| Solubility | DMSO : < 1 mg/mL (ultrasonic;warming;heat to 66°C) (insoluble or slightly soluble) ∣ Ethanol : < 1 mg/mL (ultrasonic;warming) (insoluble) |
| Molecular Weight (g/mol) | 534.39 g/mol |
| CAS | 158013-42-4 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?