missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mesothelin Antibody (MSLN/6443R), Novus Biologicals™
Rabbit Monoclonal Antibody
2875.00 NOK - 5120.00 NOK
Specifications
| Antigen | Mesothelin |
|---|---|
| Clone | MSLN/6443R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18697274
|
Novus Biologicals
NBP3-20433-100ug |
100 μg |
5120.00 NOK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18639794
|
Novus Biologicals
NBP3-20433-20ug |
20 μg |
2875.00 NOK
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mesothelin Monoclonal antibody specifically detects Mesothelin in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Mesothelin | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| 10mM PBS with 0.05% BSA | |
| 10232 | |
| IgG κ | |
| Protein A or G purified |
| MSLN/6443R | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPFSMRP, Pre-pro-megakaryocyte-potentiating factor, soluble MPF mesothelin related protein | |
| Recombinant fragment (around aa 273-407) of human Mesothelin protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title