Learn More
3M™ Mesh Visor V4J-SV
3290.00 NOK valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Use promo code "22912" to get your promotional price.
Click to view available options
Model: V4J-SV
Description
- Etched Stainless Steel Visor
- Optimal mesh design for enhanced stability and protection
- Mesh Visors are designed to fit 3M™ Hard Hats
- All mesh visors are tested and approved under the European Standard EN 371 and meet the requirements for increased robustness (S marking)
Specifications
Certifications/Compliance | CE Approved, EN 1731 |
Thickness (Metric) | 0.3 mm |
Product Type | Mesh Visor |
For Use With (Application) | Forestry |
Model | V4J-SV |
No. per Pack | 10/Pk. |
Material | Etched Stainless Steel Mesh |