Learn More
3M™ Mesh Visor 5C
1205.00 NOK valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Use promo code "22912" to get your promotional price.
Click to view available options
Material: Stainless Steel Mesh
Description
- Stainless Steel visor with a matt black finish
- Excellent stability, even after long use
- Compatible with the 3M™ V5 Face Shield system for mounting on a 3M™ Hard Hats Series G3000 or G22
- Snap in features makes it quick and easy to combine the face shield to the face shield holder
Specifications
Certifications/Compliance | CE Approved, EN 1731 |
Product Type | Mesh Visor |
For Use With (Equipment) | V5 Face Shield System |
For Use With (Application) | Forestry |
Material | Stainless Steel Mesh |