missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MERS-CoV Spike Protein Antibody, mFluor Violet 610 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
MERS-CoV Spike Protein Polyclonal antibody specifically detects MERS-CoV Spike Protein in MERS-CoV samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | MERS-CoV Spike Protein |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 610 SE |
| Dilution | Western Blot, ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | MERS S glycoprotein, MERS S1, MERS S2, MERS Spike glycoprotein, MERS Spike Protein, MERS-CoV S glycoprotein, Middle East respiratory syndrome-related coronavirus Spike glycoprotein, Spike glycoprotein, spike protein, Spike RBD, surface glycoprotein |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, a synthetic peptide corresponding to the N-terminus of the S1 subunit of MERS-CoV Spike Protein |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?