Learn More
Mercury(Ii) Chloride, ACS Reagent, Honeywell Fluka™
Product Code. 15602000 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. 15602000
Brand: Honeywell-Fluka 2154655G
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
ACS Reagent,≥99.5%
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7487-94-7
Molecular Formula Cl2Hg
Molecular Weight (g/mol) 271.49
MDL Number MFCD00011041
InChI Key LWJROJCJINYWOX-UHFFFAOYSA-L
Synonym mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2
PubChem CID 24085
ChEBI CHEBI:31823
IUPAC Name dichloromercury
SMILES [Cl-].[Cl-].[Hg++]
Specifications
Quantity 5 g
Linear Formula HgCl2
UN Number 1624
Solubility Information Soluble in diethyl ether, acetic acid, ethyl acetate, glycerol, alcohol, acetone (salt form), water
Formula Weight 271.5g/mol
Percent Purity ≥99.5%
Grade ACS Reagent
Vapor Pressure 1.3mmHg (236°C)
Chemical Name or Material Mercury(Ii) Chloride