Learn More
Mercury ICP Standard, Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
Honeywell Fluka™ analytical standards offers a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. Our products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
Specifications
Specifications
| Chemical Name or Material | Mercury ICP Standard |
| Packaging | Glass Bottle |
| CAS | 10045-94-0 |
| Technique | ICP-OES and ICP-MS |
| pH | <2 |
| Melting Point | 79°C |
| Quantity | 100 mL |
| Grade | ICP |