missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mer Antibody (MERTK/3024) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14269JF525
This item is not returnable.
View return policy
Description
Mer Monoclonal antibody specifically detects Mer in Human samples. It is validated for Immunohistochemistry (Paraffin),Protein Array
Specifications
| Mer | |
| Monoclonal | |
| Janelia Fluor 525 | |
| c-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer | |
| Recombinant human Mer protein fragment (around aa 55-148) (exact sequence is proprietary) (Uniprot: Q12866) | |
| 0.1 mL | |
| Cancer, Phospho Specific, Protein Kinase, Signal Transduction, Vision | |
| 10461 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Immunohistochemistry (Paraffin), Protein Array | |
| MERTK/3024 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction