Learn More
Mequitazine Sulfoxide N-Oxide (Mequitazine N,S-Dioxide), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Mequitazine Sulfoxide N-Oxide (Mequitazine N,S-Dioxide)
Specifications
Specifications
| Chemical Name or Material | Mequitazine Sulfoxide N-Oxide |
| CAS | 62705-13-9 |
| Analyte or Component Names | Mequitazine Sulfoxide N-Oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H22 N2 O2 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | 10-[(1-Oxido-1-azabicyclo[2.2.2]oct-3-yl)methyl]-10H-phenothiazine 5-Oxide, 10H-Phenothiazine, 10-(1-azabicyclo[2.2.2]oct-3-ylmethyl)-, N,5-dioxide, Mequitazine Sulfoxide N-Oxide |
| InChI Formula | InChI=1S/C20H22N2O2S/c23-22-11-9-15(10-12-22)16(14-22)13-21-17-5-1-3-7-19(17)25(24)20-8-4-2-6-18(20)21/h1-8,15-16H,9-14H2 |
| SMILES | [O-][N+]12CCC(CC1)C(CN3c4ccccc4S(=O)c5ccccc35)C2 |
| IUPAC Name | 10-[(1-oxido-1-azoniabicyclo[2.2.2]octan-3-yl)methyl]phenothiazine 5-oxide |
| Show More |