Learn More
Mepyramine Maleate, Mikromol™
Description
Mepyramine Maleate
Specifications
Specifications
| Chemical Name or Material | Mepyramine maleate |
| CAS | 59-33-6 |
| Analyte or Component Names | Mepyramine maleate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H23 N3 O . C4 H4 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Mepyramine Maleate, N1-[(4-Methoxyphenyl)methyl]-N2,N2-dimethyl-N1-2-pyridinyl-1,2-ethanediamine (2Z)-2-butenedioate (1:1), N-[(4-Methoxyphenyl)methyl]-N',N'-dimethyl-N-2-pyridinyl-1,2-ethanediamine (2Z)-2-butenedioate (1:1), N-[(4-Methoxyphenyl)methyl]-N',N'-dimethyl-N-2-pyridinyl-1,2-ethanediamine (Z)-2-butenedioate (1:1), 2-[(2-Dimethylaminoethyl)(p-methoxybenzyl)amino]pyridine maleate, 2-[[2-(Dimethylamino)ethyl](p-methoxybenzyl)amino]pyridine Bimaleate, AH, Anisopyradamine, Antamine, Anthisan maleate, Antihist, Antisan, Diaminide maleate, Enrumay, Histalet Forte, Histalon, Histan, Histapyran, Histatex, Histavet P, Mepyramine hydrogen maleinate, Mepyramine maleate, Minihist, NSC 3604, Neo-Antergan maleate, Neoantergan maleate, Paramal, Paraminyl, Paraminyl maleate, Parmal, Prefrin A, Pymafed, Pyra Maleate, Pyramal, Pyranilamine maleate, Pyraninyl, Pyranisamine maleate, Pyrilamine maleate, Renstamin, Stamine, Stangen, Stangen maleate, Statomin maleate, Thylogen, Thylogen maleate, 1,2-Ethanediamine, N-[(4-methoxyphenyl)methyl]-N',N'-dimethyl-N-2-pyridinyl-, (2Z)-2-butenedioate (1:1) (9CI), 1,2-Ethanediamine, N-[(4-methoxyphenyl)methyl]-N',N'-dimethyl-N-2-pyridinyl-, (Z)-2-butenedioate (1:1), Pyridine, 2-[[2-(dimethylamino)ethyl](p-methoxybenzyl)amino]-, maleate (1:1) (8CI), 2-[(2-Dimethylaminoethyl)(p-methoxybenzyl)amino]-pyridine maleate, N-p-Methoxybenzyl-N',N'-dimethyl-N-alpha-pyridylethylenediamine maleate |
| InChI Formula | InChI=1S/C17H23N3O.C4H4O4/c1-19(2)12-13-20(17-6-4-5-11-18-17)14-15-7-9-16(21-3)10-8-15;5-3(6)1-2-4(7)8/h4-11H,12-14H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1- |
| SMILES | COc1ccc(CN(CCN(C)C)c2ccccn2)cc1.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;N'-[(4-methoxyphenyl)methyl]-N,N-dimethyl-N'-pyridin-2-ylethane-1,2-diamine |
| Show More |