Learn More
Mepyramine maleate, MedChemExpress
Product Code. 16458716
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458716 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16458716 Supplier Medchem Express Supplier No. HYB1281
 Restricted Item
Estimated Shipment: 27-11-2026
MedChemExpress Mepyramine maleate, a first generation antihistamine, is an antagonist of histamine H1 receptor, with Kds of 0.8 nM, 5200 nM and >3000 nM for H1, H2, and H3 receptor, respectively, and a pKd of 9.4 for H1 receptor.
Mepyramine maleate, a first generation antihistamine, is an antagonist of histamine H1 receptor, with Kds of 0.8 nM, 5200 nM and >3000 nM for H1, H2, and H3 receptor, respectively, and a pKd of 9.4 for H1 receptor.
Specifications
Chemical Name or Material Mepyramine maleate
CAS 59-33-6
Color White
Purity Grade Notes Research
Molecular Formula C21H27N3O5
Quantity 100 mg
Synonym Pyrilamine maleate
Solubility Information DMSO : ≥ 100 mg/mL (249.09 mM) ∣H2O : 50 mg/mL (124.55 mM; Need ultrasonic)
SMILES CN(C)CCN(CC1=CC=C(OC)C=C1)C2=NC=CC=C2.O=C(O)/C=C\C(O)=O
Molecular Weight (g/mol) 401.46
Formula Weight 401.46
Percent Purity 99.96%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation