Learn More
Trajan Scientific and Medical™ MEPS Syringes for Agilent GCs
Miniaturization of conventional sample preparation devices. Trajan Scientific and Medical MEPS Syringes for Agilent GCs are micro extraction by packed sorbent.
Brand:  Trajan Scientific and Medical™ 005292
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15248483
  • 1910.00 NOK / Each
Estimated Shipment: 02-07-2024
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • For use with Agilent GCs
Specifications
Specifications
54.1 mm
100 μL
Agilent and Shimadzu MEPS Syringe