Learn More
Mepivacaine Hydrochloride, TRC
Product Code. 30135759
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135759 1 g 1g
30133372 250 mg 250mg
17949719 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135759 Supplier Toronto Research Chemicals Supplier No. TRCM2250601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1722-62-9
Molecular Formula C15 H22 N2 O . Cl H
Molecular Weight (g/mol) 282.81
Synonym Mepivacaine Hydrochloride, N-(2,6-Dimethylphenyl)-1-methyl-2-piperidinecarboxamide hydrochloride (1:1), N-(2,6-Dimethylphenyl)-1-methyl-2-piperidinecarboxamide monohydrochloride;1-Methyl-2',6'-pipecoloxylidide monohydrochloride, (±)-Mepivacaine hydrochloride, Carbocaina, Carbocaine hydrochloride, Carbocaine monohydrochloride, Chlorocain, Meaverin, Mepicaton, Mepident, Mepivacaine hydrochloride, Mepivacaine monohydrochloride, Mepivastesin, Optocain, Polocaine, Scandicain, Scandonest
IUPAC Name N-(2,6-dimethylphenyl)-1-methylpiperidine-2-carboxamide;hydrochloride
SMILES Cl.CN1CCCCC1C(=O)Nc2c(C)cccc2C
Specifications
Quantity 1 g
InChI Formula InChI=1S/C15H22N2O.ClH/c1-11-7-6-8-12(2)14(11)16-15(18)13-9-4-5-10-17(13)3;/h6-8,13H,4-5,9-10H2,1-3H3,(H,16,18);1H
Formula Weight 282.1499
Percent Purity >95%
Chemical Name or Material Mepivacaine Hydrochloride
Grade HPLC