Learn More
Mepivacaine hydrochloride, MedChemExpress
Product Code. 16490968
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490968 5 g 5g
16452209 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16490968 Supplier Medchem Express Supplier No. HYB0517A
MedChemExpress Mepivacaine hydrochloride binds to specific voltage-gated sodium ion channels in neuronal cell membranes, which inhibits both sodium influx and membrane depolarization.
Mepivacaine hydrochloride binds to specific voltage-gated sodium ion channels in neuronal cell membranes, which inhibits both sodium influx and membrane depolarization.
Specifications
Chemical Name or Material Mepivacaine hydrochloride
CAS 1722-62-9
Color White
Purity Grade Notes Research
Molecular Formula C15H23ClN2O
Quantity 5 g
Solubility Information H2O : 100 mg/mL (353.59 mM; Need ultrasonic)
SMILES O=C(C(CCCC1)N1C)NC(C(C)=CC=C2)=C2C.Cl
Molecular Weight (g/mol) 282.81
Formula Weight 282.81
Percent Purity 99.83%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation