missing translation for 'onlineSavingsMsg'
Learn More

MEPCE Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11526-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16848663

  • 3828.53 NOK / 150µL
Estimated Shipment: 12-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

S-adenosyl-L-methionine-dependent methyltransferase that adds a methylphosphate cap at the 5′-end of 7SK snRNA, leading to stabilize it.
Specifications

Specifications

MEPCE
Polyclonal
Unconjugated
MEPCE
Bin3 homolog, MEPCE, methylphosphate capping enzyme
Rabbit
Antigen Affinity Chromatography
RUO
231803, 304361, 561384, 56257
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A3KQ55, Q7L2J0, Q8K3A9
Mepce
MEPCE Fusion Protein Ag2092
150 μL
Primary
Human, Mouse, Rat, Zebrafish
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers