Learn More
Epredia™ Menzel™ Microscope Coverslips
Product Code. 11961988 Shop All Epredia Products
Change view
Click to view available options
Length (Metric):
18 mm
20 mm
24 mm
Width (Metric):
18 mm
20 mm
24 mm
Unit Size:
Pack of 1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Width (Metric) Unit Size
11961988 20 mm 20 mm Pack of 1000
15767572 18 mm 18 mm Pack of 1000
15707592 24 mm 24 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11961988 Supplier Epredia™ Supplier No. BB02000200A153MNZ0
  • ONLINE EXCLUSIVE PRICE
    411.34 NOK
    210.62 NOK / Pack of 1000
    Save 200.72 NOK
    48% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Specifications
Product Type Coverslip
Shape Square
Material Borosilicate Glass
Length (Metric) 20 mm
Width (Metric) 20 mm
Thickness (Metric) #1 (0.13 to 0.16 mm)
Dimensions (L x W) 20 x 20 mm
For Use With (Equipment) Automated equipment
Quantity 1000/Pk.