Learn More
Uvex™ Men Softshell 7431-Yellow
Product Code. p-7232012 Shop All Uvex Products
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
Unit Size:
Each
Product Code. 17206671
Brand: Uvex™ 8938213
Only null left
Add to basket
Add to basket
All that is essential is that everyone can be seen despite the poor visibility. The high-visibility clothing from the uvex protection flash collection uses bright reflective strips. And of course the cargo trousers and softshell jackets with their optimum fit can also be seen in daylight
  • Softshell jacket - uvex protection flash model-no.: 7431
  • Material: 100% polyester, with membrane in 3-layer construction, approx. 240 gsm
  • Windproof and water-repellent high vis softshell jacket
  • One chest pocket left with zip and two big side pockets with zip, lightweight and although breathable and hard-wearing material, 10.000 mm water column rating
  • Sizes: XS - 6XL
  • Oversize surcharge: from size XL+10%
  • Care instructions: 60°C washable, chlorinate impossible, tumble dryer: I point, ironing: I point, dry cleaning: P certified according to: EN ISO 20471 X2
TRUSTED_SUSTAINABILITY
Specifications
Size 2X-Large
Gender Men's
Material Polyester
Color Yellow, Gray
Closure Type Zipper
Disposable Reusable
Product Type Hi-vis Jacket
Thickness 285 g/m²