Learn More
Alexandra™ Men's Anti-Pill Fleece Jacket, Navy
Description
- Two zip hip pockets
- Elasticated drawcord with stopper
- Unisex
- Up to 40° wash
- Back length: 72 - 74cm (increases with size)
- Fabric Weight: 300gsm
- 100% polyester
Specifications
Specifications
| Size | Large |
| Gender | Unisex |
| Material | Polyester |
| Color | Navy |
| Style | NG19 |
| Weight (Metric) | 300 gsm |