Learn More
Mettler Toledo™ Membrane Kit
Product Code. 10798473 Shop All Mettler Toledo Products
Change view
Click to view available options
Includes:
4 Membrane Bodies, 1 O-ring Set, 25 mL Electrolyte
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
10798473 4 Membrane Bodies, 1 O-ring Set, 25 mL Electrolyte 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10798473 Supplier Mettler Toledo™ Supplier No. 52200024
  • ONLINE EXCLUSIVE PRICE
    14660.00 NOK
    7846.52 NOK / 1 sets
    Save 6813.48 NOK
    46% Off
 Restricted Item
Estimated Shipment: 27-10-2026
Used as an accessory part for the InPro O2 sensor family. Mettler Toledo™ Membrane Kit for the the InPro O2 sensor enables reliable inline measurement of dissolved oxygen in a wide spectrum of processes that operate under sterile/hygienic conditions.
Outstanding features of the O2 sensors are:
  • hygienic design, capable of withstanding CIP procedures
  • stainless steel surfaces with highly polished finish
  • steam sterilizable and autoclavable up to 140°C
  • low detection limit at high signal stability
  • O-rings with FDA and USP Class VI approval
  • minimal maintenance requirement
  • ISM (Intelligent Sensor Management)
TRUSTED_SUSTAINABILITY
Specifications
Type Mettler Toledo Membrane Kit
Includes 4 Membrane Bodies, 1 O-ring Set, 25 mL Electrolyte
compliance-icons
Product Identifier
  • O2 electrolyte
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: potassium hydroxide