Learn More
Memantine-d6 Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Memantine-d6 Hydrochloride |
| CAS | 1189713-18-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C12 2H6 H15 N . Cl H |
| Quantity | 10 mg |
| Synonym | 3,5-Di(methyl-d3)-tricyclo[3.3.1.13,7]decan-1-amine Hydrochloride (1:1), Memantin-d6-Dimethyl |
| InChI Formula | InChI=1S/C12H21N.ClH/c1-10-3-9-4-11(2,6-10)8-12(13,5-9)7-10;/h9H,3-8,13H2,1-2H3;1H/i1D3,2D3; |
| SMILES | Cl.[2H]C([2H])([2H])C12CC3CC(N)(CC(C3)(C1)C([2H])([2H])[2H])C2 |
| IUPAC Name | 3,5-bis(trideuteriomethyl)adamantan-1-amine;hydrochloride |
| Molecular Weight (g/mol) | 221.8 |
| Show More |