Learn More
Gibco™ MEM Vitamin Solution (100X)
GREENER_CHOICE
Product Code. 11550386
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. Quantity unitSize
11550386 100 mL 100mL
1 options
Product Code. 11550386
Brand: Gibco™ 11120037
MEM Vitamin Solution (100X)
Gibco™ MEM Vitamin Solution is used as a growth supplement for cell culture medium, to increase cell growth and viability. Gibco™ MEM Vitamin Solution is formulated to contain 100x the vitamins found in the standard Minimum Essential Medium (MEM). The complete formulation is available.
Product Use
For in vitro diagnostic use.
Dual-site cGMP Manufacturing and Quality System
Gibco™ MEM Vitamin Solution is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ MEM Vitamin Solution product made in our Grand Island facility (11120-052). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage –5 to –20°C
Description MEM Vitamin Solution, 100mL, Fewer Resources Used, Less Waste, Used as a Growth Supplement for Cell Culture Medium, to Increase Cell Growth and Viability
Product Type MEM Vitamin Solution
For Use With (Application) Mammalian Cell Culture
Quantity 100 mL
Regulatory Status RUO - research use only