Learn More
Melatonin-d7, TRC
Specifications
Specifications
| Chemical Name or Material | Melatonin-d7 |
| CAS | 615251-68-8 |
| Molecular Formula | C13 2H7 H9 N2 O2 |
| Quantity | 10 mg |
| Synonym | Acetamide-2,2,2-d3, N-[2-(5-methoxy-1H-indol-3-yl)ethyl-1,1,2,2-d4]- (9CI, ACI), N-[2-(5-Methoxy-1H-indol-3-yl)ethyl-1,1,2,2-d4]acetamide-2,2,2-d3 (ACI), N-(Acetyl-d3)-5-methoxytryptamine-α,α,β,β-d4, Melatonin-d7, Melatonin-D7 (acetamidoethyl-D7) |
| InChI Formula | InChI=1S/C13H16N2O2/c1-9(16)14-6-5-10-8-15-13-4-3-11(17-2)7-12(10)13/h3-4,7-8,15H,5-6H2,1-2H3,(H,14,16)/i1D3,5D2,6D2 |
| SMILES | [2H]C([2H])([2H])C(=O)NC([2H])([2H])C([2H])([2H])c1c[nH]c2ccc(OC)cc12 |
| IUPAC Name | 2,2,2-trideuterio-N-[1,1,2,2-tetradeuterio-2-(5-methoxy-1H-indol-3-yl)ethyl]acetamide |
| Molecular Weight (g/mol) | 239.32 |
| Formula Weight | 239.1651 |