missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melatonin, 98.5%, MP Biomedicals™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
Color | White to Beige |
Quantity | 1 g |
Formula Weight | 232.3 |
Percent Purity | 0.985 |
Physical Form | Powder |
Chemical Name or Material | Melatonin |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'