Learn More
Brand™ Melamine Formaldehyde Mortar with Pestle
Product Code. 10038210 Shop All Brand Products
Change view
Click to view available options
Diameter (Metric) Outer:
125mm
150mm
Height (Metric):
80mm
90mm
Unit Size:
Each
Product Code. Diameter (Metric) Outer Height (Metric) unitSize
10038210 150mm 90mm Each
10495031 125mm 80mm Each
2 options
Product Code. 10038210
Brand: Brand™ 110710
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Resistant to boiling and autoclavable (at 121°C).
  • White in color
  • With foot and spout
  • Provided individually
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Diameter (Metric) Outer 150mm
Product Type Mortar and Pestle
Material Melamine Formaldehyde
Height (Metric) 90mm