Learn More
BrandTech™ VITLAB™ Melamine Formaldehyde Instrument Tray
Support your laboratory instruments, tools, and sensitive utensils using the VITLAB™ Melamine Formaldehyde Instrument Tray, which is stable and self-supporting.
Brand:  BrandTech™ 72198
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10493222
  • 910.00 NOK / Pack of 5
Estimated Shipment: 03-06-2024
Description
  • White Melamine
  • Flat shape
  • Rounded corners
  • Smooth surface
  • Easy to clean
Specifications
Specifications
290 x 160 x 35 mm
35 mm
Polyurethane
160 mm
Rectangular
White
290 mm
Instrument Tray
5/Pk.