Learn More
3M™ Melamine Floor Pads, Gray/White
Product Code. 17188610
Click to view available options
Diameter (Metric):
330 mm
370 mm
406 mm
432 mm
505 mm
Unit Size:
Pack of 5
Product Code. 17188610
Brand: 3M™ 7000062727
Only null left
Add to basket
Add to basket
Specially designed for use on stone, concrete, VCT, marble, and other porous floor surfaces. These highly effective pads make light work of ingrained dirt, and unsightly scuff marks. Environmentally preferable since no cleaning additives or chemicals required.
  • Floor pad for modern micro-porous floor surfaces such as ceramic, terracotta, marble, and concrete
  • Cleans most dirt and marks with water alone, no cleaning additives or chemicals required
  • Environmentally preferable
  • Cleans up to 1000 m2 for single disc cleaning machines and 10,000 m2 for scrubber driers
TRUSTED_SUSTAINABILITY
Specifications
Product Type Floor Pads
Color Gray, White
Diameter (Metric) 370 mm
For Use With (Application) Cleaning
For Use With (Equipment) Porous Floor Surfaces