Learn More
3M™ Melamine Floor Pads, Gray/White
Product Code. 17178610
Change view
Click to view available options
Diameter (Metric):
330 mm
370 mm
406 mm
432 mm
505 mm
Unit Size:
Pack of 5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
17178610 330 mm Pack of 5
17188610 370 mm Pack of 5
17198610 406 mm Pack of 5
17138570 432 mm Pack of 5
17138640 505 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17178610 Supplier 3M™ Supplier No. 7000062726
  • ONLINE EXCLUSIVE PRICE
    4030.00 NOK
    2538.93 NOK / Pack of 5
    Save 1491.07 NOK
    36% Off
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
Specially designed for use on stone, concrete, VCT, marble, and other porous floor surfaces. These highly effective pads make light work of ingrained dirt, and unsightly scuff marks. Environmentally preferable since no cleaning additives or chemicals required.
  • Floor pad for modern micro-porous floor surfaces such as ceramic, terracotta, marble, and concrete
  • Cleans most dirt and marks with water alone, no cleaning additives or chemicals required
  • Environmentally preferable
  • Cleans up to 1000 m2 for single disc cleaning machines and 10,000 m2 for scrubber driers
TRUSTED_SUSTAINABILITY
Specifications
Product Type Floor Pads
Color Gray, White
Diameter (Metric) 330 mm
For Use With (Application) Cleaning
For Use With (Equipment) Porous Floor Surfaces