Learn More
Usbeck Laborgeraete™ Meker Burner
Specifications
Specifications
| Product Type | Meker Burner |
| Gas Type | Butane, LPG, Natural Gas |
| Diameter (Metric) | 32 mm (Head) |
| Diameter (Metric) Outer Mixing Tube | 28 mm |
| Height (Metric) | 200 mm |
| Max. Temperature (Metric) | 1490°C |
| No. per Pack | 1 Ea. |
| Weight (Metric) | 335 g |