Learn More
MEK6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12745-1-AP-20UL
 View more versions of this product
Product Code. 16850624
  • 1710.33 NOK / 20µL
Description
This gene encodes a member of the dual specificity protein kinase family, which functions as a mitogen-activated protein (MAP) kinase kinase. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals. This protein phosphorylates and activates p38 MAP kinase in response to inflammatory cytokines or environmental stress. As an essential component of p38 MAP kinase mediated signal transduction pathway, this gene is involved in many cellular processes such as stress induced cell cycle arrest, transcription activation and apoptosis.
Specifications
Specifications
MEK6
Polyclonal
Unconjugated
MAP2K6
MAP kinase kinase 6, MAP2K6, MAPK/ERK kinase 6, MAPKK 6, MAPKK6, MEK 6, MEK6, MKK6, PRKMK6, SAPKK3
Rabbit
Antigen Affinity Chromatography
RUO
114495, 26399, 5608
-20°C
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P52564, P70236
Map2k6
MEK6 Fusion Protein Ag3706
20 μL
Primary
Human, Rat, Mouse