missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MAP2K5 is a dual specificity protein kinase that belongs to the MAP kinase kinase family. It specifically interacts with and activates MAPK7/ERK5. This kinase itself can be phosphorylated and activated by MAP3K3/MEKK3, as well as by atypical protein kinase C isoforms (aPKCs). The signal cascade mediated by MAP2K5 is involved in growth factor stimulated cell proliferation and muscle cell differentiation.
Specifications
Specifications
| Antigen | MEK5 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.13 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | MAP2K5 |
| Gene Accession No. | Q13163, Q62862, Q9WVS7 |
| Gene Alias | HsT17454, MAP kinase kinase 5, MAP2K5, MAPK/ERK kinase 5, MAPKK 5, MAPKK5, MEK 5, MEK5, MKK5, PRKMK5 |
| Gene Symbols | Map2k5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?