Learn More
MEK2 Mouse anti-Human, Unconjugated, Clone: 1H8C2, Proteintech
Product Code. 16810026 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16810026 150 μL 150µL
16820026 20 μL 20µL
2 options
Product Code. 16810026
Brand: Proteintech 674101IG150UL
Mouse Monoclonal Antibody
The MEK2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MEK2. This antibody recognizes human, mouse, rat antigen. The MEK2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Antigen MEK2
Applications Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
Classification Monoclonal
Clone 1H8C2
Concentration 1000 μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 5605
Target Species Human, Rat, Mouse
Isotype IgG1