missing translation for 'onlineSavingsMsg'
Learn More

MEK1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 51080-1-AP-20UL

 View more versions of this product

Product Code. 16856375

  • 1710.33 NOK / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is specific to MEK1.

MEK1/mitogen activated protein kinase/ERK kinase 1/microtubule-associated protein 2 kinase is a dual specificity kinase and catalyses both a threonine and a tyrosine residue on MAP kinases ERK1 and ERK2. Mek1 is activated by phosphorylation of serine 218 and 222 residues by Raf1. It is known to be involved in the signaling during stress activate response, apoptosis and proliferative induction by cytokines. Mutations in the gene can lead to cardiofaciocutaneous syndrome.
Specifications

Specifications

MEK1
Polyclonal
Unconjugated
MAP2K1
ERK activator kinase 1, MAP kinase kinase 1, MAP2K1, MAPK/ERK kinase 1, MAPKK 1, MAPKK1, MEK 1, MEK1, MKK1, PRKMK1
Rabbit
Antigen Affinity Chromatography
RUO
5604
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q02750
MAP2K1
Peptide
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.