Learn More
STEMCELL Technologies MegaCult™-C Complete Kit with Cytokines
Product Code. 17118251
Encompass
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17118251 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17118251 Supplier STEMCELL Technologies Supplier No. 04971
Complete kit including collagen, media, staining kit and chamber slides for human CFU-Mk assays
  • MegaCult™-C Complete Kit with Cytokines includes all reagents required for the optimal growth of megakaryocyte progenitors (CFU-Mk) within double chamber slides and for the detection of CFU-Mk colonies by immunocytochemical staining
  • MegaCult™-C medium is optimized for the growth of CFU-Mk in human bone marrow, mobilized peripheral blood and cord blood samples
  • It is suitable for use with CD34+-enriched cells, mononuclear cells and cells isolated by other purification methods
Specifications
Cell Type Hematopoietic Stem and Progenitor Cells
For Use With (Application) Cell Culture, Colony Assay, Functional Assay
Quantity 1 Kit