Learn More
Micronova™ MegaClean™ Heavy-Duty Cleaner
Product Code. 13157736 Shop All Micronova Products
Change view
Click to view available options
Volume (English):
1 qt.
1 gal.
5 gal.
Unit Size:
15.14 L
19 L
7.6 L
Product Code. Volume (English) unitSize
13157736 1 qt. 7.6 L
11932475 1 gal. 15.14 L
13137736 5 gal. 19 L
3 options
Product Code. 13157736
Brand: Micronova™ MC1Q
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A non-flammable, heavy duty cleaner and degreaser that removes grease, adhesives and photo-resist stains from most surfaces. Available in 1 or 5 gallon bottles and a 1 quart spray bottle.
The Micronova MegaClean™ Heavy-Duty Cleaner offers an effective alternative to toxic, flammable, solvent-based cleaners, such as acetone and isopropyl alcohol. Filtered to 0.1 microns, this cleaner is ideal for semiconductor, electronics, aerospace and clean-build operations.
  • Non-flammable, heavy duty cleanroom degreaser
  • Removes grease, adhesives and photo-resist stains from most surfaces
  • Ideal for semiconductor, electronics, aerospace and clean-build operations
  • Filtered to 0.1 microns
  • Available in 1 or 5 gallon refill bottles and a 1 quart spray bottle
  • Biodegradable
  • Water soluble
  • Effective at low concentrations
Specifications
Product Type MegaClean Cleaner
Color-coded No
Handle Type None
Sterility Non-sterile
Certifications/Compliance Non-flammable, biodegradeable
Description Heavy Duty Detergent, Degreaser
Filtered Filtered
For Use With (Application) An effective alternative to toxic and flammable solvent based cleaners
For Use With (Equipment) Mops and Buckets or Wipes and Sponge
Volume (English) 1 qt.
Volume (Metric) 0.9 L