Learn More
Medroxy Progesterone 17-Acetate, TRC
Product Code. 18020720
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18020720 50 mg 50mg
17989959 10 g 10g
17999749 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18020720 Supplier Toronto Research Chemicals Supplier No. TRCM20356050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 71-58-9
Molecular Formula C24 H34 O4
Molecular Weight (g/mol) 386.52
Synonym 6alpha-Methyl-3,20-dioxopregn-4-en-17-yl acetate, Medroxyprogesterone acetate, Megestrol Acetate Imp. A (EP), Pregn-4-ene-3,20-dione, 17-(acetyloxy)-6-methyl-, (6α)-, Pregn-4-ene-3,20-dione, 17-hydroxy-6α-methyl-, acetate (7CI,8CI), Progesterone, 17-hydroxy-6α-methyl-, acetate (6CI), (6α)-17-(Acetyloxy)-6-methylpregn-4-ene-3,20-dione, 17α-Acetoxy-6α-methylpregn-4-ene-3,20-dione, 17α-Acetoxy-6α-methylprogesterone, 17α-Hydroxy-6α-methyl-4-pregnene-3,20-dione 17-acetate, 17α-Hydroxy-6α-methylpregn-4-ene-3,20-dione acetate, 17α-Hydroxy-6α-methylprogesterone acetate, 6α-Methyl-17-acetoxyprogesterone, 6α-Methyl-17R-hydroxyprogesterone acetate, 6α-Methyl-17α-Hydroxy-4-pregnene-3,20-dione acetate, 6α-Methyl-17α-acetoxy-δ4-pregnene-3,20-dione, 6α-Methyl-17α-acetoxypregn-4-ene-3,20-dione, 6α-Methyl-17α-hydroxyprogesterone acetate, Amen, Aragest, Aragest 5, Asconale, CBP 1011, Clinofem, Clinovir, Colirest, Curretab, Cycrin, DMPA, Depo-Clinovir, Depo-Prodasone, Depo-Provera, Depo-Ralovera, Depo-SubQ Provera 104, Depo-progestin, Depo-promone, Depomedroxyprogesterone acetate, Depot-medroxyprogesterone acetate, Farlutal, Farlutin, G-Farlutal, Gestapuran, Hematrol, Hysron, Indivina, Lutopolar, Lutoral, MAP, MAP (steroid), MPA, MPA Gyn 5, Med-Pro, Medroprogesterone acetate, Medroxyprogesterone 17-acetate, Meprate, Metigestrona, NSC 21171, NSC 26386, Nadigest, Nidaxin, Novo-Medrone, Oragest, Perlutex, Perlutex Leo, Prempack C, Prodasone, Progespon, Progestalfa, Progeston, Progevera, Provera, Proverone, Ralovera, Repromap, Repromix, Sirprogen, Sodelut G, U 8839, Veramix
IUPAC Name [(6S,8R,9S,10R,13S,14S,17R)-17-acetyl-6,10,13-trimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate
SMILES C[C@H]1C[C@@H]2[C@H](CC[C@@]3(C)[C@H]2CC[C@]3(OC(=O)C)C(=O)C)[C@@]4(C)CCC(=O)C=C14
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C24H34O4/c1-14-12-18-19(22(4)9-6-17(27)13-21(14)22)7-10-23(5)20(18)8-11-24(23,15(2)25)28-16(3)26/h13-14,18-20H,6-12H2,1-5H3/t14-,18+,19-,20-,22+,23-,24-/m0/s1
Formula Weight 386.2457
Percent Purity >95%
Chemical Name or Material Medroxyprogesterone acetate
Grade HPLC