Learn More
CHEMGENE MedLab Multi-Surface Disinfectant - Packaging in Norwegian, Finnish, Swedish, Danish
Shop All Chemgene ProductsDescription
- CHEMGENE MedLab is a range of surface disinfectants created specifically to meet the demands of laboratory environments
- These Multi-Surface Disinfectants are proven to be effective under the following microbiological test conditions; Bactericidal: EN 17387, EN 13727 Legionella: EN 13623. Yeasticidal: EN 13624, EN 17387. Fungicidal: EN 17387, EN 13624 Virucidal: EN 14476, EN 16777 (Vaccinia virus, Norovirus, Adenovirus) Bacteriophage: EN 13610
- Proven efficacy against resistant pathogens
- Denatures DNA/RNA
- Biodegradable formulation
- Available in eucalyptus scent or un-fragranced
- Manufactured in the UK to ISO 9001 quality standards
- Non-corrosive in-use
- Alcohol-free
- Suitable for autoclaving
- No rinsing required
- Compatible with a wide range of materials including mild steel, stainless steel, copper, aluminium, acetal, nylon and rubber.
- Bottle made from 30% recycled HDPE
- Bottle and cap recyclable after use
Specifications
Specifications
| Product Type | Multi-purpose Cleaner |
| Color-coded | No |
| Form | Concentrated Liquid |
| pH | 10.5 to 11.4 |
| Scent | Unscented |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| Color | Clear |
| Dimensions (L x W x H) | 133 x 185 x 282 mm |
| Disposable | Limited-use |
| Show More |
Use biocides safely. Always read the product label and information before use.