Learn More
CHEMGENE MedLab Multi-Surface Disinfectant - Packaging in Norwegian, Finnish, Swedish, Danish
Product Code. 31451497 Shop All Chemgene Products
Change view
Click to view available options
Color:
Blue
Clear
Scent:
Eucalyptus
Unscented
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Scent
31451497 Blue Eucalyptus
31451500 Clear Unscented
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
SAFETY PRECAUTIONS DANGER: Causes skin irritation. Causes serious eye damage. Very toxic to aquatic life with long lasting effects. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wash contaminated skin thoroughly after handling. IF ON SKIN: Wash with plenty of soap and water. Dispose of contents/container in accordance with national regulations.
Product Code. 31451497 Supplier CHEMGENE Supplier No. SKU077D
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
 Restricted Item
Estimated Shipment: 16-11-2026
SAFETY PRECAUTIONS DANGER: Causes skin irritation. Causes serious eye damage. Very toxic to aquatic life with long lasting effects. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wash contaminated skin thoroughly after handling. IF ON SKIN: Wash with plenty of soap and water. Dispose of contents/container in accordance with national regulations.
Fast-acting multi-surface disinfectant for labs, critical services, and forensic settings; decontaminates surfaces and denatures DNA/RNA.
  • CHEMGENE MedLab is a range of surface disinfectants created specifically to meet the demands of laboratory environments
  • These Multi-Surface Disinfectants are proven to be effective under the following microbiological test conditions; Bactericidal: EN 17387, EN 13727 Legionella: EN 13623. Yeasticidal: EN 13624, EN 17387. Fungicidal: EN 17387, EN 13624 Virucidal: EN 14476, EN 16777 (Vaccinia virus, Norovirus, Adenovirus) Bacteriophage: EN 13610
  • Proven efficacy against resistant pathogens
  • Denatures DNA/RNA
  • Biodegradable formulation
  • Available in eucalyptus scent or un-fragranced
  • Manufactured in the UK to ISO 9001 quality standards
  • Non-corrosive in-use
  • Alcohol-free
  • Suitable for autoclaving
  • No rinsing required
  • Compatible with a wide range of materials including mild steel, stainless steel, copper, aluminium, acetal, nylon and rubber.
  • Bottle made from 30% recycled HDPE
  • Bottle and cap recyclable after use
Specifications
Product Type Multi-purpose Cleaner
Color-coded No
Form Concentrated Liquid
pH 10.5 to 11.4
Scent Eucalyptus
Sterility Non-sterile
Autoclavable Autoclavable
Color Blue
Dimensions (L x W x H) 133 x 185 x 282 mm
Disposable Limited-use
Filtered Filtered
Flammable No
Height (Metric) 282 mm
Packaging Bottle - 30% recycled HDPE
Volume (Metric) 5 L
Width (Metric) 185 mm
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • MedLab Multi Surface Disinfectant Concentrate 5
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash contaminated skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Alkyl (C12-16) dimethylbenzyl ammonium chloride, Didecyldimethylammonium chloride, Limonene
Use biocides safely. Always read the product label and information before use.