Learn More
Lenz Laborglasinstrumente™ Medium Shape Weighing Bottles
Product Code. 11886164
Change view
Click to view available options
Diameter (Metric):
30 mm
38 mm
48 mm
75 mm
Unit Size:
Each
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
11886164 75 mm Each
11798104 48 mm Pack of 6
11788104 30 mm Pack of 6
10030181 38 mm Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11886164 Supplier Lenz Laborglasinstrumente™ Supplier No. 05565580
  • ONLINE EXCLUSIVE PRICE
    704.00 NOK
    579.58 NOK / Pack of 1
    Save 124.42 NOK
    17% Off
Estimated Shipment: 14-12-2026
Specifications
Includes Interchangeable NS stopper, inscription label
Quantity 1Pack
Stopper Number NS 80/12
Product Line Bottle
Diameter (Metric) 75 mm
Product Type Weighing Bottle