Learn More
PAL International™ Medipal™ Medical Disinfectant Wipes
Product Code. 15824165
Change view
Click to view available options
Dimensions (L x W):
300 x 200 mm
Unit Size:
Pack of 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
15824165 300 x 200 mm Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15824165 Supplier PAL International™ Supplier No. S522110MPCE
  • ONLINE EXCLUSIVE PRICE
    83.45 NOK
    79.60 NOK / Pack of 200
    Save 3.85 NOK
    4% Off
 Restricted Item
Estimated Shipment: 28-12-2026
Supports healthcare institutions to achieve both their clinical and budgetary targets. Using Medipal™ wipes will help infection prevention teams to fight infection and ensure healthy, clinically clean environments for patients.
  • They contain a powerful dual biocidal solution, highly effective from 30 seconds against a wide range of pathogenic microorganisms and are intended for cleaning and disinfecting surfaces of non invasive medical devices
  • Medipal™ Disinfectant Wipes are bactericidal (including TB), virucidal, yeasticidal and fungicidal
  • They are tested to a range of EN standards including EN13727, EN14476, EN14348, EN14561, EN14563
  • They are alcohol free and gentle on surfaces, making them suitable for everyday use
  • They can be used in combination with Medipal High Level Disinfectant Wipes in areas of particular high risk
  • Low foaming & low residue
Specifications
Composition Didecyldimethylammonium chloride (DDAC) & Polyhexamethylene biguanide hydrochloride (PHMB)
Container Type Soft Pack
Description Easy to transport and dispense, these wipes are suitable for general everyday cleaning and disinfection.
Dimensions (L x W) 300 x 200 mm
Quantity 200 Wipes/Pk.
compliance-icons
Product Identifier
  • Medipal 3in1 Disinfectant Wipes
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-aminoethanol, didecyldimethylammoniumchloride, N-(3-aminopropyl)-N-dodecylpropane-1,3-diamine, al