Learn More
Medical Viscosity Standard, 4.0 mPa.s (25.00°C ), Paragon Scientific™
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
- Fully traceable to international standards
- Certified in accordance to ISO 17025 & ISO 17034 under Paragon's UKAS accreditation
- Certified in strict accordance with ASTM D2162
- Tamper evident security packaging
- 12 month shelf life
- Manufactured in the United Kingdom
- Contains antibacterial agent
Specifications
Specifications
| Certifications/Compliance | ISO 17025, ISO 17034 |
| Quantity | 100 mL |
Nominal values quoted. Certified Values will vary from batch to batch.