Learn More
DWK Life Sciences Wheaton™ Media Lab Bottle, Non-Graduated, Amber
Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
125 mL
Unit Size:
Pack of 48
Description
- Manufactured from borosilicate glass which provides the best chemical resistance
- Amber borosilicate glass bottle provides protection from UV light
- Special pour lip facilitates pouring
- Packaged in partitioned carton to help prevent scratch marks on bottles
- Black phenolic screw cap can be purchased separately
Specifications
Specifications
| Capacity (Metric) | 125 mL |
| Color | Amber |
| Material | Borosilicate Glass (Type I) |
| Quantity | 48 EA |
| Product Type | Media Bottle |
| Closure Type | Not Included |