missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15951-1-AP-20UL
This item is not returnable.
View return policy
Description
The Mediator is a multiprotein coactivator that is required by DNA-binding transcription factors for activation of polymerase II (see MIM 180660)-transcribed genes. MED4 appears to be a core Mediator subunit and is found in nearly all Mediator preparations (summary by Sato et al., 2004).Specifications
| MED4 | |
| Polyclonal | |
| Unconjugated | |
| MED4 | |
| ARC36, DRIP36, HSPC126, MED4, mediator complex subunit 4, RP11 90M2.2, TRAP36, VDRIP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 29079, 306030, 67381 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q561Q8, Q9CQA5, Q9NPJ6 | |
| MED4 | |
| MED4 Fusion Protein Ag8629 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction