missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MECR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14932-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The mitochondrial trans-2-enoyl-CoA reductase (MECR), was initially identified as nuclear receptor-binding factor 1 (NRBF1), which can interact with a multitude of nuclear hormone receptors in the presence of the respective ligands. MECR has been shown to be part of the mitochondrial fatty acid synthesis (FAS II) system and to catalyze the NAPDH-dependent reduction of 2-enoyl thioesters, generating saturated acyl-groups. Overexpression of this gene in transgenic mice can lead to cardiac abnormalities, suggesting that inappropriate expression of genes of FAS II can result in the development of hereditary cardiomyopathy.Specifications
MECR | |
Polyclonal | |
Unconjugated | |
MECR | |
CGI 63, FASN2B, HsNrbf 1, MECR, NBRF1, NRBF 1, NRBF1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26922, 29470, 51102 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BV79, Q9DCS3, Q9Z311 | |
MECR | |
MECR Fusion Protein Ag6738 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |