missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MeCP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
2005.00 NOK - 3235.00 NOK
Specifications
| Antigen | MeCP2 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 2 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18019793
|
Novus Biologicals
NB100-56326SS |
0.025 mg |
2005.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18795213
|
Novus Biologicals
NB100-56326 |
0.1 mg |
3235.00 NOK
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MeCP2 Polyclonal specifically detects MeCP2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| MeCP2 | |
| Western Blot 2 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| P51608 | |
| 4204 | |
| This antibody was generated by immunizing rabbits with a mixture of synthetic peptides corresponding to amino acids 11-25 and 181-195 of human MeCP2. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Chromatin Research, Neuroscience | |
| PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
| AUTSX3, DKFZp686A24160, MeCp-2 protein, mental retardation, X-linked 79, methyl CpG binding protein 2 (Rett syndrome), X-linked 16 | |
| MECP2 | |
| IgG | |
| Protein G purified | |
| In HeLa, a 53 kDa band is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title