Learn More
BINDER™ Mechanical and Electrical Membrane Pump VP 4
Binder™ Mechanical and Electrical Membrane Pump VP 4 with separator and emission condenser.
Brand:  BINDER™ 5013-0049
Product Code. 15621556
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Nominal air flow 1.9m³/hour
  • Final pressure 12 mbar
  • 1 phase, 230 V AC, 50 Hz