Learn More
Mebhydrolin Napadisilate, LoGiCal
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
missing translation for 'unitSize'
10mg
Description
Mebhydrolin Napadisilate
Specifications
Specifications
| Chemical Name or Material | Mebhydrolin Napadisilate |
| CAS | 6153-33-9 |
| Analyte or Component Names | Mebhydrolin Napadisilate |
| Shipping Condition | Room Temperature |
| Molecular Formula | 2 C19 H20 N2 . C10 H8 O6 S2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1,5-Naphthalenedisulfonic acid, compd. with 2,3,4,5-tetrahydro-2-methyl-5-(phenylmethyl)-1 H-pyrido[4,3-b]indole (1:2), 1,5-Naphthalenedisulfonic acid, compd. with 5-benzyl-2,3,4,5-tetrahydro-2-methyl-1 H-pyrido[4,3-b]indole (1:2) (8 CI), 2 H-Pyrido[4,3-b]indole, 5-benzyl-1,3,4,5-tetrahydro-2-methyl-, 1,5-naphthalenedisulfonate (2:1) (6 CI), 1 H-Pyrido[4,3-b]indole, 2,3,4,5-tetrahydro-2-methyl-5-(phenylmethyl)-, 1,5-naphthalenedisulfonate (2:1) (9 CI), 1 H-Pyrido[4,3-b]indole, 5-benzyl-2,3,4,5-tetrahydro-2-methyl-, 1,5-naphthalenedisulfonate (2:1) (8 CI), Diazolin, Diazoline, Fabahistin, Incidal, Mebhydrolin napadisylate, Omeril |
| InChI Formula | InChI=1 S/2C19H20N2.C10H8O6S2/c2*1-20-12-11-19-17(14-20)16-9-5-6-10-18(16)21(19)13-15-7-3-2-4-8-15;11-17(12,13)9-5-1-3-7-8(9)4-2-6-10(7)18(14,15)16/h2*2-10 H,11-14H2,1H3;1-6 H,(H,11,12,13)(H,14,15,16) |
| SMILES | CN1CCc2c(C1)c3ccccc3n2Cc4ccccc4.CN5CCc6c(C5)c7ccccc7n6Cc8ccccc8.OS(=O)(=O)c9cccc%10 c(cccc9%10)S(=O)(=O)O |
| IUPAC Name | 5-benzyl-2-methyl-3,4-dihydro-1 H-pyrido[4,3-b]indole;naphthalene-1,5-disulfonic acid |
| Show More |