Learn More
DWK Life Sciences DURAN™ Plain Cylinder, with round base, without graduation
Cylinders without graduation can be used to mix components.
Brand: DWK Life Sciences 213993407
2389.25 NOK valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 1.70000kg
Description
The multitude of different work steps in the laboratory environment demands a multitude of universally applicable glassware. These glass devices usually have a very simple shape and are characterized by their versatility.
- Plain ground rim
- With round base
Specifications
Borosilicate Glass (3.3) | |
190 mL | |
10/Pk. |
40 mm | |
Diluting solutions; Mixing several components | |
Yes |